Assay Details
| Gene Symbol: SLC22A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631352.3 | XP_005631409.1 | 7 - 8 | 879 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A18| Gene Symbol: SLC22A18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631352.3 | XP_005631409.1 | 7 - 8 | 879 | 138 | |
Set Membership: |
|