Assay Details
| Gene Symbol: SMARCB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_543533.6 | XP_543533.2 | 3 - 4 | 456 | 64 | |
| XM_852177.5 | XP_857270.2 | 3 - 4 | 433 | 64 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMARCB1
Genomic Map