Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287075.1 | NP_001274004.1 | 2 - 3 | 186 | 148 | |
| XM_005632414.3 | XP_005632471.1 | 2 - 3 | 230 | 148 | ||
| XM_014121736.2 | XP_013977211.1 | 2 - 3 | 341 | 148 | ||
| XM_022406239.1 | XP_022261947.1 | 6 - 7 | 890 | 148 | ||
| XM_022406240.1 | XP_022261948.1 | 5 - 6 | 804 | 148 | ||
| XM_022406241.1 | XP_022261949.1 | 4 - 5 | 616 | 148 | ||
| XM_022406242.1 | XP_022261950.1 | 5 - 6 | 702 | 148 | ||
More Information
Set Membership: |
|

WNT5A
Genomic Map