Assay Details
| Gene Symbol: MZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616002.3 | XP_005616059.1 | 2 - 3 | 861 | 166 | |
| XM_005616003.3 | XP_005616060.1 | 2 - 3 | 647 | 166 | ||
| XM_005616004.3 | XP_005616061.1 | 2 - 3 | 764 | 166 | ||
| XM_005616005.3 | XP_005616062.1 | 2 - 3 | 561 | 166 | ||
| XM_005616006.3 | XP_005616063.1 | 2 - 3 | 556 | 166 | ||
| XM_014114465.2 | XP_013969940.1 | 2 - 3 | 623 | 166 | ||
| XM_022426730.1 | XP_022282438.1 | 2 - 3 | 617 | 166 | ||
More Information
Set Membership: |
|

MZF1
Genomic Map