Assay Details
| Gene Symbol: GPR68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005623744.3 | XP_005623801.1 | 3 | 865 | 80 | |
| XM_005623745.3 | XP_005623802.1 | 2 | 3291 | 80 | ||
| XM_014116131.2 | XP_013971606.1 | 2 | 1880 | 80 | ||
| XM_014116132.2 | XP_013971607.1 | 2 | 530 | 80 | ||
| XM_022422333.1 | XP_022278041.1 | 2 | 1262 | 80 | ||
More Information
Set Membership: |
|

GPR68
Genomic Map