Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115577.2 | XP_013971052.1 | 5 | 1611 | 159 | |
| XM_022403823.1 | XP_022259531.1 | 4 | 1527 | 159 | ||
| XM_022403826.1 | XP_022259534.1 | 4 | 2304 | 159 | ||
More Information
Set Membership: |
|

ZNF527
Genomic Map