Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639659.3 | XP_005639716.1 | 872 | 154 | ||
| XM_005639660.3 | XP_005639717.1 | 728 | 154 | |||
| XM_014110329.2 | XP_013965804.1 | 746 | 154 | |||
| XM_022414131.1 | XP_022269839.1 | 725 | 154 | |||
| XM_843160.5 | XP_848253.2 | 844 | 154 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

OTULIN
Genomic Map