Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434310.4 | XP_003434358.1 | 4 | 332 | 85 | |
| XM_545783.6 | XP_545783.2 | 5 | 499 | 252 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434310.4 | XP_003434358.1 | 4 | 332 | 85 | |
| XM_545783.6 | XP_545783.2 | 5 | 499 | 252 | ||
Set Membership: |
|