Assay Details
| Gene Symbol: C1QTNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417946.1 | XP_022273654.1 | 1 | 353 | 79 | |
| XM_022417947.1 | XP_022273655.1 | 1 | 353 | 79 | ||
| XM_848980.5 | XP_854073.1 | 1 | 352 | 79 | ||
More Information
Set Membership: |
|

C1QTNF3
Genomic Map