Assay Details
| Gene Symbol: CALHM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639996.3 | XP_003640044.1 | 2 | 565 | 79 | |
| XM_014109127.2 | XP_013964602.1 | 2 | 1173 | 79 | ||
| XM_022411853.1 | XP_022267561.1 | 2 | 523 | 79 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CALHM2
Genomic Map