Assay Details
| Gene Symbol: MIEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625860.2 | XP_005625917.1 | 2 - 3 | 341 | 143 | |
| XM_005625861.2 | XP_005625918.1 | 2 - 3 | 283 | 143 | ||
| XM_847396.3 | XP_852489.1 | 2 - 3 | 344 | 143 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MIEF1
Genomic Map