Assay Details
| Gene Symbol: ADPGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638573.3 | XP_005638630.1 | 7 | 1247 | 98 | |
| XM_005638574.3 | XP_005638631.1 | 7 | 1244 | 98 | ||
More Information
Set Membership: |
|

ADPGK
Genomic Map