Assay Details
| Gene Symbol: OTUD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641335.3 | XP_005641392.1 | 3 - 4 | 1546 | 143 | |
| XM_014111601.2 | XP_013967076.1 | 3 - 4 | 1544 | 143 | ||
| XM_538031.6 | XP_538031.1 | 3 - 4 | 752 | 143 | ||
More Information
Set Membership: |
|

OTUD5
Genomic Map