Assay Details
| Gene Symbol: ZNF106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638308.3 | XP_005638365.1 | 8 - 9 | 3533 | 164 | |
| XM_014109449.2 | XP_013964924.1 | 6 - 7 | 3384 | 164 | ||
More Information
Set Membership: |
|

ZNF106
Genomic Map