Assay Details
| Gene Symbol: PIGO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626748.3 | XP_005626805.2 | 9 - 10 | 3257 | 100 | |
| XM_005626749.3 | XP_005626806.2 | 9 - 10 | 3254 | 100 | ||
| XM_005626750.3 | XP_005626807.1 | 9 - 10 | 3422 | 100 | ||
| XM_005626751.3 | XP_005626808.1 | 9 - 10 | 3394 | 100 | ||
| XM_005626752.3 | XP_005626809.1 | 9 - 10 | 3397 | 100 | ||
| XM_022425545.1 | XP_022281253.1 | 10 - 11 | 2642 | 100 | ||
| XM_531985.6 | XP_531985.3 | 9 - 10 | 3182 | 100 | ||
More Information
Set Membership: |
|

PIGO
Genomic Map