Assay Details
| Gene Symbol: HOMER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416761.1 | XP_022272469.1 | 1009 | 77 | ||
| XM_022416762.1 | XP_022272470.1 | 1071 | 77 | |||
| XM_844616.5 | XP_849709.1 | 1693 | 77 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HOMER1
Genomic Map