Assay Details
| Gene Symbol: CHST9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421508.1 | XP_022277216.1 | 5 | 1080 | 127 | |
| XM_022421509.1 | XP_022277217.1 | 4 | 1041 | 127 | ||
| XM_022421510.1 | XP_022277218.1 | 5 | 970 | 127 | ||
More Information
Set Membership: |
|

CHST9
Genomic Map