Assay Details
| Gene Symbol: POLDIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625794.3 | XP_005625851.1 | 2 | 328 | 195 | |
| XM_531709.6 | XP_531709.3 | 2 | 328 | 195 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLDIP3| Gene Symbol: POLDIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625794.3 | XP_005625851.1 | 2 | 328 | 195 | |
| XM_531709.6 | XP_531709.3 | 2 | 328 | 195 | ||
Set Membership: |
|