Assay Details
| Gene Symbol: KCNH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639256.4 | XP_003639304.1 | 1163 | 153 | ||
| XM_014116400.2 | XP_013971875.1 | 1163 | 153 | |||
| XM_022422821.1 | XP_022278529.1 | 3074 | 153 | |||
More Information
Set Membership: |
|

KCNH6
Genomic Map