Assay Details
| Gene Symbol: ANXA8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417626.1 | XP_022273334.1 | 11 - 12 | 1038 | 246 | |
| XM_022417627.1 | XP_022273335.1 | 11 - 12 | 1027 | 246 | ||
| XM_536412.6 | XP_536412.2 | 10 - 11 | 935 | 246 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANXA8L1
Genomic Map