Assay Details
| Gene Symbol: LOC487628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109562.2 | XP_013965037.1 | 8 - 9 | 1396 | 131 | |
| XM_022412902.1 | XP_022268610.1 | 8 - 9 | 1379 | 131 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC487628
Genomic Map