Assay Details
| Gene Symbol: FLAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622776.3 | XP_005622833.1 | 3 | 932 | 146 | |
| XM_022421322.1 | XP_022277030.1 | 3 | 970 | 146 | ||
| XM_849970.5 | XP_855063.1 | 3 | 970 | 146 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FLAD1
Genomic Map