Assay Details
| Gene Symbol: RELA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631472.3 | XP_005631529.1 | 3 - 4 | 286 | 97 | |
| XM_005631473.2 | XP_005631530.1 | 3 - 4 | 333 | 97 | ||
| XM_005631474.3 | XP_005631531.1 | 3 - 4 | 311 | 97 | ||
| XM_014121307.2 | XP_013976782.1 | 3 - 4 | 285 | 97 | ||
| XM_022405652.1 | XP_022261360.1 | 3 - 4 | 316 | 97 | ||
| XM_540850.6 | XP_540850.2 | 3 - 4 | 308 | 97 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RELA
Genomic Map