Assay Details
| Gene Symbol: RAB3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625620.3 | XP_005625677.1 | 939 | 183 | ||
| XM_005625621.2 | XP_005625678.1 | 1043 | 183 | |||
| XM_005625622.3 | XP_005625679.1 | 1039 | 183 | |||
| XM_014117228.2 | XP_013972703.1 | 845 | 183 | |||
| XM_022424198.1 | XP_022279906.1 | 940 | 183 | |||
| XM_022424199.1 | XP_022279907.1 | 940 | 183 | |||
| XM_538280.5 | XP_538280.4 | 1043 | 183 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RAB3IP
Genomic Map