Assay Details
| Gene Symbol: KIAA1217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617001.2 | XP_005617058.1 | 3421 | 134 | ||
| XM_014121726.2 | XP_013977201.1 | 3681 | 134 | |||
| XM_014121729.2 | XP_013977204.1 | 3598 | 134 | |||
| XM_014121732.2 | XP_013977207.1 | 3599 | 134 | |||
| XM_014121735.2 | XP_013977210.1 | 3745 | 134 | |||
| XM_014121742.2 | XP_013977217.1 | 3491 | 134 | |||
| XM_014121744.2 | XP_013977219.1 | 3492 | 134 | |||
| XM_014121749.1 | XP_013977224.1 | 3313 | 134 | |||
| XM_014121750.2 | XP_013977225.1 | 3155 | 134 | |||
| XM_014121759.2 | XP_013977234.1 | 3029 | 134 | |||
| XM_014121760.2 | XP_013977235.1 | 2932 | 134 | |||
| XM_014121766.2 | XP_013977241.1 | 2666 | 134 | |||
| XM_014121769.2 | XP_013977244.1 | 2812 | 134 | |||
| XM_014121773.2 | XP_013977248.1 | 2595 | 134 | |||
| XM_014121778.1 | XP_013977253.1 | 3316 | 134 | |||
| XM_022406473.1 | XP_022262181.1 | 3180 | 134 | |||
More Information
Set Membership: |
|

KIAA1217
Genomic Map