Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005642053.3 | XP_005642110.1 | 5 | 913 | 147 | |
| XM_005642054.3 | XP_005642111.1 | 5 | 913 | 147 | ||
| XM_022416390.1 | XP_022272098.1 | 6 | 976 | 147 | ||
| XM_022416391.1 | XP_022272099.1 | 6 | 973 | 147 | ||
| XM_848556.5 | XP_853649.1 | 5 | 913 | 147 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMLHE
Genomic Map