Assay Details
| Gene Symbol: KCNMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003299.1 | NP_001003299.1 | 2 | 213 | 162 | |
| XM_005618870.3 | XP_005618927.1 | 3 | 653 | 162 | ||
| GenBank mRNA | U41002.1 | - | 2 | 213 | 162 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

KCNMB1
Genomic Map