Assay Details
| Gene Symbol: C9H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_848743.4 | XP_853836.3 | 9 - 10 | 1038 | 177 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H17orf75| Gene Symbol: C9H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_848743.4 | XP_853836.3 | 9 - 10 | 1038 | 177 | |
Set Membership: |
|