Assay Details
| Gene Symbol: LOC100855730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121697.2 | XP_013977172.1 | 929 | 150 | ||
| XM_014121698.2 | XP_013977173.1 | 798 | 150 | |||
| XM_014121699.2 | XP_013977174.1 | 783 | 150 | |||
| XM_022406292.1 | XP_022262000.1 | 678 | 150 | |||
More Information
Set Membership: |
|

LOC100855730
Genomic Map