Assay Details
| Gene Symbol: HVCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636220.3 | XP_005636277.1 | 1007 | 189 | ||
| XM_005636222.3 | XP_005636279.1 | 881 | 189 | |||
| XM_005636224.3 | XP_005636281.1 | 1110 | 189 | |||
| XM_005636225.3 | XP_005636282.1 | 1115 | 189 | |||
| XM_005636226.3 | XP_005636283.1 | 1070 | 189 | |||
| XM_014107901.2 | XP_013963376.1 | 972 | 189 | |||
| XM_022410333.1 | XP_022266041.1 | 936 | 189 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HVCN1
Genomic Map