Assay Details
| Gene Symbol: TWNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637625.3 | XP_005637682.1 | 4 - 5 | 912 | 165 | |
| XM_022412113.1 | XP_022267821.1 | 4 - 5 | 2440 | 165 | ||
| XM_022412114.1 | XP_022267822.1 | 5 - 6 | 2314 | 165 | ||
| XM_022412115.1 | XP_022267823.1 | 6 - 7 | 951 | 165 | ||
| XM_543974.5 | XP_543974.1 | 4 - 5 | 2389 | 165 | ||
More Information
Set Membership: |
|

TWNK
Genomic Map