Assay Details
| Gene Symbol: RNF145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619242.2 | XP_005619299.1 | 10 - 11 | 1772 | 122 | |
| XM_546272.6 | XP_546272.2 | 10 - 11 | 1763 | 122 | ||
More Information
Set Membership: |
|

RNF145
Genomic Map