Assay Details
| Gene Symbol: PACRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418714.1 | XP_022274422.1 | 408 | 119 | ||
| XM_022418715.1 | XP_022274423.1 | 406 | 119 | |||
| XM_022418717.1 | XP_022274425.1 | 405 | 119 | |||
| XM_541188.6 | XP_541188.2 | 403 | 119 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PACRG
Genomic Map