Assay Details
| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120523.2 | XP_013975998.1 | 4 | 448 | 89 | |
| XM_540195.6 | XP_540195.2 | 5 | 496 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM150A| Gene Symbol: TMEM150A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120523.2 | XP_013975998.1 | 4 | 448 | 89 | |
| XM_540195.6 | XP_540195.2 | 5 | 496 | 89 | ||
Set Membership: |
|