Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403481.1 | XP_022259189.1 | 6 - 7 | 1488 | 172 | |
| XM_532684.6 | XP_532684.2 | 10 - 11 | 2355 | 172 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PRMT9
Genomic Map