Assay Details
| Gene Symbol: DLA-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011723.1 | NP_001011723.1 | 3 | 399 | 188 | |
| XM_005627066.2 | XP_005627123.1 | 3 | 497 | 188 | ||
| XM_005627067.2 | XP_005627124.1 | 3 | 497 | 188 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DLA-DRA
Genomic Map