Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617344.3 | XP_005617401.1 | 9 - 10 | 2212 | 133 | |
| XM_005617345.3 | XP_005617402.1 | 9 - 10 | 2215 | 133 | ||
| XM_022409104.1 | XP_022264812.1 | 9 - 10 | 2216 | 133 | ||
| XM_853486.5 | XP_858579.1 | 9 - 10 | 2200 | 133 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SH3RF2
Genomic Map