Assay Details
| Gene Symbol: UQCRH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629036.3 | XP_005629093.1 | 3 | 224 | 146 | |
| XM_014119566.2 | XP_013975041.1 | 4 | 339 | 294 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014119566.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UQCRH
Genomic Map