Assay Details
| Gene Symbol: CREBZF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633413.2 | XP_005633470.1 | 1 | 919 | 143 | |
| XM_014122761.2 | XP_013978236.1 | 1 | 919 | 143 | ||
| XM_014122762.2 | XP_013978237.1 | 1 | 919 | 143 | ||
| XM_014122763.2 | XP_013978238.1 | 1 | 919 | 143 | ||
| XM_014122764.2 | XP_013978239.1 | 1 | 919 | 143 | ||
| XM_014122765.2 | XP_013978240.1 | 1 | 919 | 143 | ||
| XM_014122766.2 | XP_013978241.1 | 1 | 919 | 143 | ||
| XM_014122767.2 | XP_013978242.1 | 1 | 919 | 143 | ||
| XM_014122768.2 | XP_013978243.1 | 1 | 919 | 143 | ||
| XM_022407574.1 | XP_022263282.1 | 1 | 919 | 143 | ||
| XM_022407575.1 | XP_022263283.1 | 1 | 919 | 143 | ||
| XM_022407577.1 | XP_022263285.1 | 1 | 919 | 143 | ||
| XM_022407578.1 | XP_022263286.1 | 1 | 919 | 143 | ||
| XM_022407579.1 | XP_022263287.1 | 1 | 919 | 143 | ||
| XM_022407580.1 | XP_022263288.1 | 1 | 919 | 143 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CREBZF
Genomic Map