Assay Details
| Gene Symbol: NRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639416.4 | XP_003639464.1 | 1 | 113 | 137 | |
| XM_022425815.1 | XP_022281523.1 | 1 | 113 | 176 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NRM
Genomic Map