Assay Details
| Gene Symbol: EDN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002956.1 | NP_001002956.1 | 3 | 424 | 174 | |
| GenBank mRNA | AB115087.1 | - | 3 | 424 | 174 | |
| AF333433.1 | - | 2 | 175 | 174 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EDN1
Genomic Map