Assay Details
| Gene Symbol: STK16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640686.3 | XP_005640743.1 | 5 - 6 | 796 | 162 | |
| XM_545656.6 | XP_545656.1 | 6 - 7 | 833 | 162 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STK16
Genomic Map