Assay Details
| Gene Symbol: ERI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629093.3 | XP_005629150.1 | 4 - 5 | 819 | 120 | |
| XM_014119479.2 | XP_013974954.1 | 4 - 5 | 821 | 120 | ||
| XM_014119480.2 | XP_013974955.1 | 4 - 5 | 560 | 120 | ||
| XM_014119481.2 | XP_013974956.1 | 3 - 4 | 470 | 120 | ||
| XM_014119482.2 | XP_013974957.1 | 3 - 4 | 281 | 120 | ||
| XM_014119483.2 | XP_013974958.1 | 2 - 3 | 209 | 120 | ||
| XM_022428038.1 | XP_022283746.1 | 4 - 5 | 819 | 120 | ||
| XM_022428039.1 | XP_022283747.1 | 4 - 5 | 820 | 120 | ||
| XM_022428040.1 | XP_022283748.1 | 2 - 3 | 462 | 120 | ||
| XM_845756.4 | XP_850849.2 | 4 - 5 | 816 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014119482.2, XM_014119483.2 transcripts.
Set Membership: |
|

ERI3
Genomic Map