Assay Details
| Gene Symbol: MITF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003337.1 | NP_001003337.1 | 386 | 92 | ||
| XM_005632016.2 | XP_005632073.1 | 601 | 92 | |||
| XM_005632018.2 | XP_005632075.1 | 601 | 92 | |||
| XM_005632019.3 | XP_005632076.1 | 269 | 92 | |||
| GenBank mRNA | AY240952.1 | - | 386 | 92 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MITF
Genomic Map