Assay Details
| Gene Symbol: CRYBB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048113.1 | NP_001041578.1 | 2 - 3 | 102 | 123 | |
| GenBank mRNA | DQ324463.1 | - | 2 - 3 | 102 | 123 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CRYBB2
Genomic Map