Assay Details
| Gene Symbol: ALOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205113.1 | NP_001192042.1 | 8 - 9 | 1291 | 153 | |
| GenBank mRNA | HQ657323.1 | - | 8 - 9 | 1291 | 153 | |
More Information
Set Membership: |
|

ALOX5
Genomic Map