Assay Details
| Gene Symbol: CYP26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411974.1 | XP_022267682.1 | 3 - 4 | 5096 | 57 | |
| XM_844281.5 | XP_849374.1 | 2 - 3 | 5238 | 57 | ||
More Information
Set Membership: |
|

CYP26A1
Genomic Map