Assay Details
| Gene Symbol: CREB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628667.3 | XP_005628724.1 | 7 - 8 | 824 | 124 | |
| XM_014119045.2 | XP_013974520.1 | 8 - 9 | 894 | 124 | ||
| XM_022427521.1 | XP_022283229.1 | 7 - 8 | 1318 | 124 | ||
| XM_022427522.1 | XP_022283230.1 | 7 - 8 | 1316 | 124 | ||
| XM_022427523.1 | XP_022283231.1 | 8 - 9 | 937 | 124 | ||
| XM_022427524.1 | XP_022283232.1 | 7 - 8 | 1438 | 124 | ||
| XM_022427525.1 | XP_022283233.1 | 7 - 8 | 1321 | 124 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CREB5
Genomic Map