Assay Details
| Gene Symbol: BCKDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627565.3 | XP_005627622.1 | 7 - 8 | 858 | 120 | |
| XM_005627566.3 | XP_005627623.1 | 7 - 8 | 860 | 120 | ||
| XM_532213.6 | XP_532213.2 | 7 - 8 | 858 | 120 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BCKDHB
Genomic Map