Assay Details
| Gene Symbol: CDKN1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627279.3 | XP_005627336.1 | 2 - 3 | 536 | 56 | |
| XM_532125.6 | XP_532125.3 | 2 - 3 | 555 | 56 | ||
| GenBank mRNA | AJ830019.1 | - | 1 - 2 | 349 | 56 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDKN1A
Genomic Map